4-Aminopiperidine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Piperidin-4-amine
43292-0050
43292-0250
TS43292-0050EA
88
USD
TS43292-0050
TS43292-0250
4-Aminopiperidine 95%
4-Aminopiperidine
Formula:
C₅H₁₂N₂ MW: 100.16 g/mol Boiling Pt: 65 °C (18Torr) Density: 0.954 g/cm³ Flash Pt: 46 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179399 CAS Number: 13035-19-3 UN: 2733 ADR: 3,III |
Specification Test Results
Appearance | Colorless to yellow to brown Liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
UV | at 260 nm A: ≤0.02 |
Titration after Ion exchange | ≥99.0 % (on dry substance) |
Membrane filtration test | (on 0.5 M soln.) Passes test |
Suitability | (HPLC gradient) Passes test |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...