2-(Methylthio)ethanol 99%

Supplier: Thermo Scientific Chemicals

21010-0050 21010-0250 21010-1000
TS21010-0050EA 28 USD
TS21010-0050 TS21010-0250 TS21010-1000
2-(Methylthio)ethanol 99%
2-(Methylthio)ethanol
Formula: C₃H₈OS
Density: 1.06 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00002908
CAS Number: 5271-38-5
EINECS: 226-090-3
UN: 3334
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4934 to 1.4954 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR