2-Chloro-p-benzoquinone 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chlorocyclohexa-2,5-diene-1,4-dione, 2-Chloro-1,4-benzoquinone
33122-0050
33122-0250
TS33122-0050EA
86
USD
TS33122-0050
TS33122-0250
2-Chloro-p-benzoquinone 95%
2-Chloro-p-benzoquinone
Formula:
C₆H₃ClO₂ MW: 142.54 g/mol Melting Pt: 52…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075260 CAS Number: 695-99-8 |
Specification Test Results
Appearance | Yellow to brown Crystals |
Infrared spectrum | Conforms |
Melting point | 52 to 57 °C |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...