2-Chloro-5-fluoropyrimidine 98%

Supplier: Thermo Scientific Chemicals
Danger

42981-0010 42981-2500
TS42981-0010EA 180 USD
TS42981-0010 TS42981-2500
2-Chloro-5-fluoropyrimidine 98%
2-Chloro-5-fluoropyrimidine
Formula: C₄H₂ClFN₂
Boiling Pt: 149…162 °C (1013hPa)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD03788197
CAS Number: 62802-42-0
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %
Arsenic (As) ≤3 ppm
Lead (Pb) ≤4 ppm
Loss on ignition ≤32.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR