Myristoyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetradecanoyl chloride

18063-0050 18063-1000 18063-5000
TS18063-0050EA 27 USD
TS18063-0050 TS18063-1000 TS18063-5000
Myristoyl chloride 97%
Myristoyl chloride
Formula: C₁₄H₂₇ClO
Boiling Pt: 172…176 °C (5mbar)
Melting Pt: –1 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000741
CAS Number: 112-64-1
EINECS: 203-994-6
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4480 to 1.4500 (20°C, 589 nm)
GC ≥96.0 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR