cis-2,3-Epoxybutane 97%

Supplier: Thermo Scientific Chemicals
Danger

30787-0010 30787-0050
TS30787-0010EA 55 USD
TS30787-0010 TS30787-0050
cis-2,3-Epoxybutane 97%
cis-2,3-Epoxybutane
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 60…61 °C
Melting Pt: 84…83 °C
Density: 0.826 g/cm³
Flash Pt: –21.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00005128
CAS Number: 1758-33-4
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3820 to 1.3840 (20°C, 589 nm)
Water ≤2.5 %
GC ≥96.0 % (corrected for H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR