3-Chlorophthalic anhydride 95+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloroisobenzofuran-1,3-dione

38129-0010 38129-0050 38129-0250
TS38129-0010EA 35 USD
TS38129-0010 TS38129-0050 TS38129-0250
3-Chlorophthalic anhydride 95+%
3-Chlorophthalic anhydride
Formula: C₈H₃ClO₃
Melting Pt: 123…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00023107
CAS Number: 117-21-5
EINECS: 204-179-8

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or flakes and/or chunks
Infrared spectrum Conforms
Melting point 122 to 126 °C
Titration by coupling ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR