2-Chloroethylammonium chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloroethylamine hydrochloride, 2-Chloroethylamine HCl, 2-Chloroethanamine hydrochloride

42286-0010 42286-0250 42286-1000
TS42286-0010EA 347 USD
TS42286-0010 TS42286-0250 TS42286-1000
2-Chloroethylammonium chloride 98%
2-Chloroethylammonium chloride
Formula: C₂H₇Cl₂N
Melting Pt: 140…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00012887
CAS Number: 870-24-6
EINECS: 212-793-2

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder, crystals or chunks
Infrared spectrum Conforms
Melting point 140 to 150 °C
Loss on drying ≤1 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR