2,6-Dimethoxy-3-pyridineboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
44492-0010
44492-0050
TS44492-0010EA
200
USD
TS44492-0010
TS44492-0050
2,6-Dimethoxy-3-pyridineboronic acid pinacol ester 97%
2,6-Dimethoxy-3-pyridineboronic acid pinacol ester
Formula:
C₁₃H₂₀BNO₄ Melting Pt: 39…44 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03788240 CAS Number: 214360-59-5 |
Specification Test Results
Appearance | Colorless to off-white Liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...