Dodecane (mixture of isomers)
Supplier: Thermo Scientific Chemicals
|
Danger
|
36577-0250
36577-0010
36577-0050
TS36577-0250EA
670
USD
TS36577-0250
TS36577-0010
TS36577-0050
Dodecane (mixture of isomers)
Dodecane (mixture of isomers)
Formula:
C₁₂H₂₆ Boiling Pt: 170…195 °C (760 mmHg) Melting Pt: –60 °C Density: 0.751 g/cm³ Flash Pt: 44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008969 CAS Number: 93685-81-5 EINECS: 297-629-8 UN: 3295 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4190 to 1.4230 (20°C, 589 nm) |
Density | (15°C) 746 to 756 kg/m3 |
Distillation range | 170 to 195 °C |
Aromatics | ≤50 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...