(±)-1-Phenyl-2-propyn-1-ol 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Phenyl-2-propyn-1-ol

39276-0010 39276-0050 39276-0250
TS39276-0010EA 35 USD
TS39276-0010 TS39276-0050 TS39276-0250
(±)-1-Phenyl-2-propyn-1-ol 98+%
(±)-1-Phenyl-2-propyn-1-ol
Formula: C₉H₈O
MW: 132.16 g/mol
Melting Pt: 22 °C
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00021860
CAS Number: 4187-87-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5530 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR