4-Amino-2-chlorophenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-2-chlorophenylboronic acid,pinacolester, 4-Amino-2-chlorophenylboronic acid, piancol ester

44448-0010 44448-0050
TS44448-0010EA 254 USD
TS44448-0010 TS44448-0050
4-Amino-2-chlorophenylboronic acid pinacol ester 97%
4-Amino-2-chlorophenylboronic acid pinacol ester
Formula: C₁₂H₁₇BClNO₂
MW: 253.54 g/mol
Melting Pt: 123…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09864184
CAS Number: 877160-63-9

Order Now

Specification Test Results

Appearance Off-white to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 120 to 125 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR