Genistein (synthetic) 99%

Supplier: Thermo Scientific Chemicals
Warning

32827-0250 32827-1000
TS32827-0250EA 129 USD
TS32827-0250 TS32827-1000
Genistein (synthetic) 99%
Genistein
Formula: C₁₅H₁₀O₅
Melting Pt: 297…298 °C
Storage Temperature: Freezer
MDL Number: MFCD00016952
CAS Number: 446-72-0
EINECS: 207-174-9

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR