Daidzein 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Daidzeol, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',7-Dihydroxyisoflavone, 7-Hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
32823-0250
32823-1000
TS32823-0250EA
114
USD
TS32823-0250
TS32823-1000
Daidzein 98%
Daidzein
Formula:
C₁₅H₁₀O₄ MW: 254.24 g/mol Boiling Pt: 513 Storage Temperature: Freezer |
MDL Number:
MFCD00016954 CAS Number: 486-66-8 EINECS: 207-635-4 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...