(5-Bromopentyl)benzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-5-phenylpentane

38474-0010 38474-0050
TS38474-0010EA 94 USD
TS38474-0010 TS38474-0050
(5-Bromopentyl)benzene 98%
(5-Bromopentyl)benzene
Formula: C₁₁H₁₅Br
MW: 227.14 g/mol
Boiling Pt: 148 °C (18 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD01075177
CAS Number: 14469-83-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5340 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR