5-Bromo-1-methylimidazole 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Bromo-1-methyl-1H-imidazole

38104-0050 38104-0250
TS38104-0050EA 154 USD
TS38104-0050 TS38104-0250
5-Bromo-1-methylimidazole 97%
5-Bromo-1-methylimidazole
Formula: C₄H₅BrN₂
MW: 161
Boiling Pt: 120 °C (15 mmHg)
Melting Pt: 40…41 °C
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD01632218
CAS Number: 1003-21-0
UN: 2923
ADR: 8 II

Order Now

Specification Test Results

Appearance White to tan Crystalline solid
Infrared spectrum Conforms
Melting point 38 to 43 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR