4-Bromo-1-methylimidazole 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromo-1-methyl-1H-imidazole
45243-0010
45243-0050
TS45243-0010EA
124
USD
TS45243-0010
TS45243-0050
4-Bromo-1-methylimidazole 95%
4-Bromo-1-methylimidazole
Formula:
C₄H₅BrN₂ MW: 161 Boiling Pt: 91…93 °C (.1hPa) Flash Pt: >110 °C Storage Temperature: Freezer |
MDL Number:
MFCD01320501 CAS Number: 25676-75-9 |
Specification Test Results
Appearance | Clear colorless to yellow to light brown Liquid |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...