2-Bromoimidazole 98%

Supplier: Thermo Scientific Chemicals
Danger

45463-0010 45463-0050
TS45463-0010EA 216 USD
TS45463-0010 TS45463-0050
2-Bromoimidazole 98%
2-Bromoimidazole
Formula: C₃H₃BrN₂
Melting Pt: 189…190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179526
CAS Number: 16681-56-4

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR