4'-Chloro-2'-hydroxyacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Chloro-2-hydroxyphenyl)ethan-1-one, 4-Chloro-2-hydroxyacetophenone

44936-0100 44936-0500
TS44936-0100EA 129 USD
TS44936-0100 TS44936-0500
4'-Chloro-2'-hydroxyacetophenone 97%
4'-Chloro-2'-hydroxyacetophenone
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Melting Pt: 27…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00238557
CAS Number: 6921-66-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline mass
Infrared spectrum Conforms
Melting point 27 to 30 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR