4-Amino-3-hydroxy-1-naphthalenesulphonic acid 90+% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Amino-2-naphthol-4-sulphonic acid, 4-Amino-3-hydroxy-α-naphthalenesulphonic acid, 4-Amino-3-hydroxynaphthalene-1-sulphonic acid, 1-Amino-2-hydroxy-4-naphthalenesulphonic acid

40085-0250 40085-1000
TS40085-0250EA 84 USD
TS40085-0250 TS40085-1000
4-Amino-3-hydroxy-1-naphthalenesulphonic acid 90+% ACS
4-Amino-3-hydroxy-1-naphthalenesulphonic acid
Formula: C₁₀H₉NO₄S
MW: 239.25 g/mol
Boiling Pt: 466 °C (1013 hPa)
Melting Pt: 295 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004019
CAS Number: 116-63-2
EINECS: 204-147-3
Merck Index: 1300453

Order Now

Specification Test Results

Appearance White to light brown-pink to light purple Powder
Infrared spectrum Conforms
Solubility (in sodium carbonate) Passes test
Residue after ignition ≤0.1 %
Titration with NaOH ≥90.0 %
Sensitivity (to phosphate) Passes test
Sulfate (SO4) ≤0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR