2-Naphthoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Naphthoyl Chloride

37659-0100 37659-0500
TS37659-0100EA 69 USD
TS37659-0100 TS37659-0500
2-Naphthoyl chloride 98%
2-Naphthoyl chloride
Formula: C₁₁H₇ClO
MW: 190.63 g/mol
Boiling Pt: 156…158 °C (7 mmHg)
Melting Pt: 49…54 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004093
CAS Number: 2243-83-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Grey to brown Crystalline chunks
Infrared spectrum Conforms
Melting point 48 to 52 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR