4-Bromothiazole 97%

Supplier: Thermo Scientific Chemicals
Warning

42945-0010 42945-0050
TS42945-0010EA 62 USD
TS42945-0010 TS42945-0050
4-Bromothiazole 97%
4-Bromothiazole
Formula: C₃H₂BrNS
Flash Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06657592
CAS Number: 34259-99-9

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR