Tris(diethylamino)phosphine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hexaethyltriaminophosphine, Hexaethylphosphorus triamide, Hexaethyl phosphorous triamide

42308-0050 42308-0250 42308-1000
TS42308-0050EA 57 USD
TS42308-0050 TS42308-0250 TS42308-1000
Tris(diethylamino)phosphine 95%
Tris(diethylamino)phosphine
Formula: C₁₂H₃₀N₃P
MW: 247.36 g/mol
Boiling Pt: 111 °C (6 mmHg)
Density: 0.89 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00009041
CAS Number: 2283-11-6
EINECS: 218-920-8
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4750 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR