5-Bromothiazole 95%

Supplier: Thermo Scientific Chemicals
Warning

42998-0010 42998-0050
TS42998-0010EA 89 USD
TS42998-0010 TS42998-0050
5-Bromothiazole 95%
5-Bromothiazole
Formula: C₃H₂BrNS
MW: 164.03 g/mol
Flash Pt: 76.7 °C
Storage Temperature: Freezer
MDL Number: MFCD07787394
CAS Number: 3034-55-7

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR