5-Bromo-2-chloropyrimidine 96%

Supplier: Thermo Scientific Chemicals
Warning

36848-0050 36848-2500
TS36848-0050EA 346 USD
TS36848-0050 TS36848-2500
5-Bromo-2-chloropyrimidine 96%
5-Bromo-2-chloropyrimidine
Formula: C₄H₂BrClN₂
Melting Pt: 78…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00483232
CAS Number: 32779-36-5

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR