5-Bromo-2,4-dichloropyrimidine 98%

Supplier: Thermo Scientific Chemicals
Danger

37136-0010 37136-0050
TS37136-0010EA 29 USD
TS37136-0010 TS37136-0050
5-Bromo-2,4-dichloropyrimidine 98%
5-Bromo-2,4-dichloropyrimidine
Formula: C₄HBrCl₂N₂
MW: 227.87 g/mol
Boiling Pt: 128 °C (15 mmHg)
Melting Pt: 29…30 °C
Density: 1.781 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00127818
CAS Number: 36082-50-5
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear to slightly hazy colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.6020 to 1.6050 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR