4-Iodopyridine 96%

Supplier: Thermo Scientific Chemicals
Warning

39129-0010 39129-0050
TS39129-0010EA 63 USD
TS39129-0010 TS39129-0050
4-Iodopyridine 96%
4-Iodopyridine
Formula: C₅H₄IN
MW: 205 g/mol
Melting Pt: 92…96 °C
Storage Temperature: Freezer
MDL Number: MFCD02093937
CAS Number: 15854-87-2

Order Now

Specification Test Results

Appearance Light brown to dark green-brown Crystals or powder (May darken on storage)
Infrared spectrum Conforms
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR