4,4-Dimethylcyclohexanone 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4,4-Dimethylcyclohexan-1-one

44881-0010 44881-0050
TS44881-0010EA 44 USD
TS44881-0010 TS44881-0050
4,4-Dimethylcyclohexanone 97%
4,4-Dimethylcyclohexanone
Formula: C₈H₁₄O
Boiling Pt: 73 °C (14 mmHg)
Melting Pt: 41…43 °C
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00234965
CAS Number: 4255-62-3
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Appearance White to tan Crystals or solid
Infrared spectrum Conforms
Melting point 41 to 45 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR