3,3,3-Trifluoropropionic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

38107-0010 38107-0050 38107-1000
TS38107-0010EA 43 USD
TS38107-0010 TS38107-0050 TS38107-1000
3,3,3-Trifluoropropionic acid 98%
3,3,3-Trifluoropropionic acid
Formula: C₃H₃F₃O₂
Boiling Pt: 146 °C (760 mmHg)
Melting Pt: 9.7 °C
Density: 1.45 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00153292
CAS Number: 2516-99-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3320 to 1.3340 (20°C, 589 nm)
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR