2-Amino-6-bromobenzothiazole 95%

Supplier: Thermo Scientific Chemicals
Warning

44229-0050 44229-0250
TS44229-0050EA 142 USD
TS44229-0050 TS44229-0250
2-Amino-6-bromobenzothiazole 95%
2-Amino-6-bromobenzothiazole
Formula: C₇H₅BrN₂S
MW: 229.1
Melting Pt: 212…214 °C
Storage Temperature: Ambient
MDL Number: MFCD00152229
CAS Number: 15864-32-1

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR