2,6-Dibromonaphthalene 99%

Supplier: Thermo Scientific Chemicals

44934-0010 44934-0050 44934-0250
TS44934-0010EA 64 USD
TS44934-0010 TS44934-0050 TS44934-0250
2,6-Dibromonaphthalene 99%
2,6-Dibromonaphthalene
Formula: C₁₀H₆Br₂
MW: 285.97 g/mol
Melting Pt: 162…167 °C
Storage Temperature: Ambient
MDL Number: MFCD01026462
CAS Number: 13720-06-4

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 162 to 167 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR