1-Butyl-3-methylimidazoliumhexafluorophosphate 98+%

Supplier: Thermo Scientific Chemicals
Warning

35417-0050 35417-0250 35417-1000
TS35417-0050EA 49 USD
TS35417-0050 TS35417-0250 TS35417-1000
1-Butyl-3-methylimidazoliumhexafluorophosphate 98+%
1-Butyl-3-methylimidazoliumhexafluorophosphate
Formula: C₈H₁₅F₆N₂P
Boiling Pt: >340 °C (760 mmHg)
Melting Pt: 6.5 °C
Density: 1.3727 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD03093295
CAS Number: 174501-64-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
HPLC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR