2-(Trifluoromethyl)benzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (2-(Trifluoromethyl)phenyl)methanamine

31295-0010 31295-0050 31295-0100
TS31295-0010EA 31 USD
TS31295-0010 TS31295-0050 TS31295-0100
2-(Trifluoromethyl)benzylamine 98%
2-(Trifluoromethyl)benzylamine
Formula: C₈H₈F₃N
MW: 175.15 g/mol
Boiling Pt: 108 °C (64 mmHg)
Density: 1.249 g/cm³
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00010297
CAS Number: 3048-01-9
EINECS: 221-258-2
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4720 (20°C, 589 nm)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR