1-(2-Bromoethyl)naphthalene 97%

Supplier: Thermo Scientific Chemicals
Warning

39956-0010 39956-0050
TS39956-0010EA 98 USD
TS39956-0010 TS39956-0050
1-(2-Bromoethyl)naphthalene 97%
1-(2-Bromoethyl)naphthalene
Formula: C₁₂H₁₁Br
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00037737
CAS Number: 13686-49-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6380 to 1.6400 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR