2-Iodopyrazine 97%

Supplier: Thermo Scientific Chemicals
Warning

39753-0010 39753-0050
TS39753-0010EA 48 USD
TS39753-0010 TS39753-0050
2-Iodopyrazine 97%
2-Iodopyrazine
Formula: C₄H₃IN₂
MW: 205.99 g/mol
Boiling Pt: 109…110 °C (34 mmHg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01319019
CAS Number: 32111-21-0

Order Now

Specification Test Results

Appearance Colorless to brown Liquid
Infrared spectrum Conforms
GC ≥96.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR