Diphenylcyclopropenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-one, 2,3-Diphenyl-2-cyclopropen-1-on, 1,2-Diphenylcyclopropen-3-one
17582-0010
17582-0050
17582-0250
TS17582-0010EA
63
USD
TS17582-0010
TS17582-0050
TS17582-0250
Diphenylcyclopropenone 98%
Diphenylcyclopropenone
Formula:
C₁₅H₁₀O MW: 206.24 g/mol Melting Pt: 117…121 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001311 CAS Number: 886-38-4 EINECS: 212-948-4 Merck Index: 1203366 |
Specification Test Results
Appearance | Beige to light orange-pink Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 117 to 121 °C |
HPLC | ≥97.5 % |
Microbial contamination | ≤5000 CFU/g (TAMC) |
pH | 6.5 to 7.5 (2 % in water) |
Chlorides | ≤2 % (as NaCl) |
Total nitrogen | 9.3 to 10.7 % |
Amino nitrogen | 1.8 to 3.2 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...