3-Bromo-4-methylthiophene 95%

Supplier: Thermo Scientific Chemicals
Warning

37996-0010 37996-0050
TS37996-0010EA 40 USD
TS37996-0010 TS37996-0050
3-Bromo-4-methylthiophene 95%
3-Bromo-4-methylthiophene
Formula: C₅H₅BrS
MW: 177.06 g/mol
Boiling Pt: 179…183 °C (770 mmHg)
Flash Pt: 78 °C
MDL Number: MFCD00130084
CAS Number: 30318-99-1
UN: 2810

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5770 to 1.5790 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR