4-Ethynylaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Aminophenyl)acetylene, 1-Ethynyl-4-aminobenzene, 1-Amino-4-ethynylbenzene, (4-Aminophenyl)ethyne

43295-0010 43295-0050
TS43295-0010EA 76 USD
TS43295-0010 TS43295-0050
4-Ethynylaniline 97%
4-Ethynylaniline
Formula: C₈H₇N
Melting Pt: 100…102 °C
Storage Temperature: Freezer
MDL Number: MFCD00168824
CAS Number: 14235-81-5

Order Now

Specification Test Results

Appearance Yellow to brown Powder
Infrared spectrum Conforms
Melting point 98 to 104 °C
GC ≥96.0 %
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.4 M soln.) Passes test
Suitability (HPLC gradient) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR