3,4-Dimethoxybenzenesulphonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
35439-0010
35439-0050
35439-0250
TS35439-0010EA
79
USD
TS35439-0010
TS35439-0050
TS35439-0250
3,4-Dimethoxybenzenesulphonyl chloride 97%
3,4-Dimethoxybenzenesulphonyl chloride
Formula:
C₈H₉ClO₄S Melting Pt: 65…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051769 CAS Number: 23095-31-0 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to beige Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 65 to 70 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...