4-Nitrobenzenesulphonyl chloride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
41585-0050
41585-0250
41585-1000
TS41585-0050EA
28
USD
TS41585-0050
TS41585-0250
TS41585-1000
4-Nitrobenzenesulphonyl chloride 95%
4-Nitrobenzenesulphonyl chloride
Formula:
C₆H₄ClNO₄S Boiling Pt: 143…144 °C (2hPa) Melting Pt: 75…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007445 CAS Number: 98-74-8 EINECS: 202-697-9 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | Yellow to brown Powder |
Infrared spectrum | Conforms |
Melting point | 75 to 80 °C |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...