3-Nitrobenzenesulphonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Nitrobenzene-1-sulphonyl chloride

41584-0250 41584-1000
TS41584-0250EA 87 USD
TS41584-0250 TS41584-1000
3-Nitrobenzenesulphonyl chloride 97%
3-Nitrobenzenesulphonyl chloride
Formula: C₆H₄ClNO₄S
Melting Pt: 60…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00007435
CAS Number: 121-51-7
EINECS: 204-476-2
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow to orange light beige Crystalline powder
Infrared spectrum Conforms
Melting point 57 to 64 °C
HPLC ≥96.0 %
Specific optical rotation -105° to -109° (20 °C, 589 nm) (C=1, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR