2-Fluorobenzenesulphonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Fluorobenzene-1-sulphonyl chloride

37578-0010 37578-0050
TS37578-0010EA 29 USD
TS37578-0010 TS37578-0050
2-Fluorobenzenesulphonyl chloride 97%
2-Fluorobenzenesulphonyl chloride
Formula: C₆H₄ClFO₂S
MW: 194.61
Boiling Pt: 242
Melting Pt: 26 °C
Density: 1.47
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042285
CAS Number: 2905-21-7
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid or solid
Infrared spectrum Conforms
Refractive index 1.5350 to 1.5390 (20°C, 589 nm)
Titration Argentometric ≥96.0 % (ex Cl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR