2-Chlorobenzenesulphonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

35746-0010 35746-0050
TS35746-0010EA 29 USD
TS35746-0010 TS35746-0050 TS35746-0250
2-Chlorobenzenesulphonyl chloride 98%
2-Chlorobenzenesulphonyl chloride
Formula: C₆H₄Cl₂O₂S
Boiling Pt: 150…152 °C (12 mmHg)
Melting Pt: 28 °C
Density: 1.548 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00051520
CAS Number: 2905-23-9
EINECS: 220-807-3
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow to orange-red Liquid or solid
Infrared spectrum Conforms
Refractive index 1.5745 to 1.5775 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR