2-Bromobenzenesulphonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

30644-0010 30644-0050
TS30644-0010EA 36 USD
TS30644-0010 TS30644-0050
2-Bromobenzenesulphonyl chloride 97%
2-Bromobenzenesulphonyl chloride
Formula: C₆H₄BrClO₂S
Boiling Pt: 98…100 °C
Melting Pt: 49…52 °C
MDL Number: MFCD00051971
CAS Number: 2905-25-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 48 to 52 °C
Titration Argentometric ≥96.0 % (ex Cl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR