1-Amino-2,2,2-trifluoroethane 99.5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2,2-Trifluoroethylamine

30350-0010 30350-0100 30350-0500
TS30350-0010EA 50 USD
TS30350-0010 TS30350-0100 TS30350-0500
1-Amino-2,2,2-trifluoroethane 99.5%
1-Amino-2,2,2-trifluoroethane
Formula: C₂H₄F₃N
MW: 99.06 g/mol
Boiling Pt: 37…38 °C (760 mmHg)
Density: 1.24 g/cm³
Flash Pt: –16 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008132
CAS Number: 753-90-2
EINECS: 212-041-3
UN: 3286
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to almost colorless Liquid
Infrared spectrum Conforms
GC ≥99.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR