Bis(4-hydroxy-N-methylanilinium)sulphate 99.0-101.5% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
p-(Methylamino)phenol sulphate, Metol, 4-Methylaminophenol sulphate, 4-(Methylamino)phenol hemisulphate, 4-Methylaminophenolsulphate, 4-(Methylamino)phenol hemisulphate salt
43444-1000
43444-5000
TS43444-1000EA
51
USD
TS43444-1000
TS43444-5000
Bis(4-hydroxy-N-methylanilinium)sulphate 99.0-101.5% ACS
4-(Methylamino)phenol sulphate
Formula:
C₁₄H₂₀N₂O₆S MW: 344.39 g/mol Melting Pt: 260 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013140 CAS Number: 55-55-0 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to off-white Crystalline powder (Discolors in air) |
Residue after ignition | ≤0.1 % |
Titration | 99.0 to 101.5 % |
Suitability | (for determ. of phosphate) Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...