Bis(4-hydroxy-N-methylanilinium)sulphate 99%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-(Methylamino)phenol sulphate, Metol, 4-Methylaminophenol sulphate, 4-(Methylamino)phenol hemisulphate, 4-Methylaminophenolsulphate, 4-(Methylamino)phenol hemisulphate salt

15680-0025 15680-0050 15680-1000 15680-5000
TS15680-0025EA 727 USD
TS15680-0025 TS15680-0050 TS15680-1000 TS15680-5000
Bis(4-hydroxy-N-methylanilinium)sulphate 99%, Extra Pure
4-(Methylamino)phenol sulphate
Formula: C₁₄H₂₀N₂O₆S
MW: 344.39 g/mol
Melting Pt: 260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013140
CAS Number: 55-55-0
EINECS: 200-237-1
UN: 2811
ADR: 6.1,III
Merck Index: 1306046

Order Now

Specification Test Results

Appearance White to beige to grey Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.3 %
Residue after ignition ≤0.15 %
Titration with NaOH ≥98.5 %
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤30 ppm
Potassium (K) ≤0.002 %
Sodium (Na) ≤0.005 %
Heavy metals (ICP-OES) ≤0.001 %
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR