Propionyl chloride 99.5% phosgene-free

Supplier: Thermo Scientific Chemicals
Danger

45897-0250 45897-5000
TS45897-0250EA 28 USD
TS45897-0250 TS45897-5000
Propionyl chloride 99.5% phosgene-free
Propionyl chloride
Formula: C₃H₅ClO
Boiling Pt: 80 °C (760 mmHg)
Melting Pt: –94 °C
Density: 1.059 g/cm³
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00000745
CAS Number: 79-03-8
UN: 1815
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4020 to 1.4050 (20°C, 589 nm)
GC ≥99.4 %
Color scale ≤20 APHA
Titration RCOCl ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR