Ethyl isothiocyanate 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
37686-0050
37686-0250
37686-1000
TS37686-0050EA
29
USD
TS37686-0050
TS37686-0250
TS37686-1000
Ethyl isothiocyanate 96%
Ethyl isothiocyanate
Formula:
C₃H₅NS MW: 87.15 g/mol Boiling Pt: 130…132 °C (760 mmHg) Melting Pt: –6 °C Density: 0.995 g/cm³ Flash Pt: 32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004820 CAS Number: 542-85-8 EINECS: 208-831-2 UN: 3286 ADR: 3,II Merck Index: 1303848 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5100 to 1.5140 (20°C, 589 nm) |
GC | ≥95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...