6-Bromoindole 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Bromo-1H-indole

39736-0010 39736-0050 39736-2500
TS39736-0010EA 162 USD
TS39736-0010 TS39736-0050 TS39736-2500
6-Bromoindole 96%
6-Bromoindole
Formula: C₈H₆BrN
MW: 196.05 g/mol
Melting Pt: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00238550
CAS Number: 52415-29-9

Order Now

Specification Test Results

Appearance White to brown Powder
Infrared spectrum Conforms
Melting point 92 to 96 °C
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR